missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Certified AR for Analysis, Fisher Chemical™
57.38 GBP valid until 2024-12-31
Use promo code "21964" to get your promotional price.
42.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
42.75 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
57.38 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "21964" to get your promotional price.
42.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
42.75 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
57.38 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Bottle
Plastic Container
Plastic-coated Glass Bottle
Description
CH4O, CAS Number-67-56-1, methyl alcohol, colonial spirit, wood naphtha, wood spirit, wood alcohol, methylol, pyroxylic spirit, methyl hydroxide, columbian spirit, carbinol, 2.5L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042
Specifications
Melting Point | -98°C |
Color | Colorless |
Boiling Point | 64.7°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 128 hPa at 20°C |
Viscosity | 0.55cp at 20°C |
Formula Weight | 32.04g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'