missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™
Click to view available options
Quantity:
6 x 1 L
Packaging:
Narrow Mouth Flint
Description
- For Use in LC/MS and UHPLC; For Laboratory, Research, or Manufacturing Use
Specifications
Melting Point | 113°C to 116°C |
Formula Weight | 32.04 |
Grade | BAKER ANALYZED™ ULTRA LC/MS |
Quantity | 6 x 1 L |
Flash Point | 12.00°C |
Packaging | Narrow Mouth Flint |
Specific Gravity | 1L = 0.79kg |
Chemical Name or Material | Methanol |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'