missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanesulfonic Acid Solution, Honeywell™ Fluka™
Click to view available options
Quantity:
10 mL
100 mL
1 L
Packaging:
Glass Bottle
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | CH3SO3H |
UN Number | UN3265 |
Beilstein | 1446024 |
Formula Weight | 96.11g/mol |
Concentration or Composition (by Analyte or Components) | approx. 1M in water |
Packaging | Glass Bottle |
Density | 1.039g/mL at 20°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'