missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10524418
101.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass Bottle
Plastic bottle
75-75-2
CH4O3S
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O
75-75-2
CH4O3S
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
6395
CHEBI:27376
methanesulfonic acid
CS(=O)(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 19.0°C
Color Yellow to Brown
Density 1.4810g/mL
Boiling Point 167.0°C (10.0 mmHg)
Flash Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3SO3H
Refractive Index 1.4252 to 1.4315
Quantity 1 L
Merck Index 15,6026
Specific Gravity 1.481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and ethanol, insoluble in hexane and methylcyclopentane, 1.50wt% benzene -0.23wt% 0-chlorotoluene (26-28°C), 0.38wt% toluene -0.47wt% ethyl disulfide (26-28°C)
Viscosity 11.6 mPa.s (25°C)
Formula Weight 96.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methanesulfonic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'