missing translation for 'onlineSavingsMsg'
Learn More

Methacryloyl chloride, 95%, contains 200 ppm MEHQ as stabilizer, AcroSeal™, Thermo Scientific™

Catalog Number 10347730
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
920-46-7
VHRYZQNGTZXDNX-UHFFFAOYSA-N
methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
13528
2-methylprop-2-enoyl chloride
CC(=C)C(=O)Cl
This item is not returnable. View return policy
920-46-7
VHRYZQNGTZXDNX-UHFFFAOYSA-N
methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
13528
2-methylprop-2-enoyl chloride
CC(=C)C(=O)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point -60.0°C
Density 1.0760g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H5ClO
Linear Formula H2C=C(CH3)COCl
Refractive Index 1.4447
Quantity 100 mL
Beilstein 02,II,394
Specific Gravity 1.076
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most common inert organic solvents,like ether,acetone and chloroform
Stabilizer 200ppm MEHQ max.
Molecular Weight (g/mol) 104.54
Formula Weight 104.54
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methacryloyl chloride, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methacryloyl chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'