missing translation for 'onlineSavingsMsg'
Learn More

Methacrylic acid, 99.5%, extra pure, stabilized, ACROS Organics™

Brand:  Acros Organics 168310010

Code : 29

Additional Details : CAS Number : 79-41-4 Weight : 1.06000kg Transport : UN number : 2531 Chem class : 1a Pack group : II


Quantity 1L
Packaging Glass Bottle

 View more versions of this product

Product Code. 10144230

  • £46.30 / 1 L


EU Stock 15
Add to basket


Specifications

Chemical Identifiers

Methacrylic acid
150-76-5
≥99.4%
C4H6O2
86.09
2-Methylpropenoic acid
Stabilized
Extra Pure
99.5%
H2C=C(CH3)CO2H
MFCD00002651

Specifications

Methacrylic acid
150-76-5
Authentic
1.4295 to 1.4315 (20°C, 589nm)
Liquid
99.5%
C4H6O2
0.2% max.
MFCD00002651
15, 6013
Clear
Stabilized
1.0100g/mL
Glass Bottle
Extra Pure
≥99.4%
250ppm (ca.) MEHQ
H2C=C(CH3)CO2H
86.09
02, 421
2-Methylpropenoic acid
1L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methacrylic acid, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyA§ thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS