missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methacryl trisepoxy cyclosiloxane

Brand:  Alfa Aesar™ 046479.22

Code : NEW

Additional Details : CAS Number : 921214-21-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15462697

  • £288.00 / 100g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Methacryl trisepoxy cyclosiloxane
C32H62O14Si4
MFCD22380702
921214-21-3
783.18
1-[3-(2-Hydroxy-3-methacryloxypropoxy)propyl]-1,3,5,7-tetramethyl-3,5,7-tris[3-(glycidyloxy)propyl]cyclotetrasiloxane

Specifications

Methacryl trisepoxy cyclosiloxane
1.11g/mL
C32H62O14Si4
MFCD22380702
Dark yellow
921214-21-3
Liquid
783.18
1-[3-(2-Hydroxy-3-methacryloxypropoxy)propyl]-1,3,5,7-tetramethyl-3,5,7-tris[3-(glycidyloxy)propyl]cyclotetrasiloxane
100g
Videos
SDS