missing translation for 'onlineSavingsMsg'
Learn More

Methacrolein, 90%, stabilized, Thermo Scientific Chemicals

Catalog Number p-156298
Click to view available options
Quantity:
10 mL
50 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
78-85-3
C4H6O
70.09
MFCD00006974
STNJBCKSHOAVAJ-UHFFFAOYSA-N
methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
6562
2-methylprop-2-enal
CC(=C)C=O
This item is not returnable. View return policy
78-85-3
C4H6O
70.09
MFCD00006974
STNJBCKSHOAVAJ-UHFFFAOYSA-N
methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
6562
2-methylprop-2-enal
CC(=C)C=O

Methacrolein, 90%, C4H6O, CAS Number-78-85-3, 2-methyl-2-propenal, 2-methylpropenal, 2-methylacrolein, 2-methylenepropanal, 2-propenal, 2-methyl, isobutenal, methacrylaldehyde, methacrylic aldehyde, methylacrylaldehyde, methacrolein, 50mL, 88% min. (GC), 01, III, 2981, 69.0 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -81.0°C
Color Colorless to Yellow
Density 0.8470g/mL
Boiling Point 69.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CHO
Refractive Index 1.4145
Quantity 10 mL
Beilstein 01, III, 2981
Specific Gravity 0.847
Solubility Information Solubility in water: 6g/100mL (20°C).
Stabilizer 0.1% hydroquinone
Viscosity 0.49 mPa.s (20°C)
Formula Weight 70.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methacrolein, 90%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'