missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacrolein, 90%, stabilized, Thermo Scientific Chemicals
102.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 mL
50 mL
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -81.0°C |
Color | Colorless to Yellow |
Density | 0.8470g/mL |
Boiling Point | 69.0°C |
Flash Point | −15°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 88% min. (GC) |
Packaging | Glass bottle |
Linear Formula | H2C=C(CH3)CHO |
Refractive Index | 1.4145 |
Show More |
Safety and Handling
Product Identifier
- Methacrolein, stabilized
- Danger
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
- H225-Highly flammable liquid and vapour.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'