missing translation for 'onlineSavingsMsg'
Learn More

Methacrolein, 90%, stabilized, ACROS Organics™

Brand:  Acros Organics 292835000

Code : 29

Additional Details : CAS Number : 78-85-3 Weight : 0.47350kg Transport : UN number : 2396 Chem class : 1 Pack group : II

Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10643572

  • £562.90 / 500mL


Estimated Shipment
20-03-2018
Add to basket

Specifications

Chemical Identifiers

Methacrolein
78-85-3
90%
H2C=C(CH3)CHO
MFCD00006974
90%
90%
C4H6O
70.09
2-Methyl-2-propenal, 2-Methylacrolein, Isobutenal, Methacrylaldehyde

Specifications

Methacrolein
78-85-3
Authentic
1.4145
0.847
90%
C4H6O
70.09
01, III, 2981
Solubility in water: 6g/100mL (20°C).
− 81.0°C
500mL
90%
0.8470g/mL
Glass bottle
Liquid
90%
0.1% hydroquinone
H2C=C(CH3)CHO
MFCD00006974
2-Methyl-2-propenal, 2-Methylacrolein, Isobutenal, Methacrylaldehyde
Clear colorless to light yellow
69.0°C
−15°C
Videos
SDS