missing translation for 'onlineSavingsMsg'
Learn More

Metaphit, Tocris Bioscience™

Acylator of PCP and σ receptors

Brand:  Tocris (Bio-Techne) 056810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13467763

  • £124.95 / 10mg

Estimated Shipment date
from Supplier 24-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acylator of PCP and σ-receptors.
Specifications

Chemical Identifiers

96316-00-6
300.464
metaphit, 1-1-3-isothiocyanatophenyl cyclohexyl piperidine, chembl41541, methaphit, tocris-0568, 1-1-3-isothiocyanatophenyl cyclohexyl pi-peridine, 1-1-3-isothiocyanato-phenyl-cyclohexyl-piperidine, piperidine, 1-1-3-isothiocyanatophenyl cyclohexyl, 1-1-3-isothiocyanato phenyl cyclohexylpiperidine
1-[1-(3-isothiocyanatophenyl)cyclohexyl]piperidine
C18H24N2S
FGSGBQAQSPSRJK-UHFFFAOYSA-N
114745
C1CCC(CC1)(C2=CC(=CC=C2)N=C=S)N3CCCCC3

Specifications

10mg
96316-00-6
metaphit, 1-1-3-isothiocyanatophenyl cyclohexyl piperidine, chembl41541, methaphit, tocris-0568, 1-1-3-isothiocyanatophenyl cyclohexyl pi-peridine, 1-1-3-isothiocyanato-phenyl-cyclohexyl-piperidine, piperidine, 1-1-3-isothiocyanatophenyl cyclohexyl, 1-1-3-isothiocyanato phenyl cyclohexylpiperidine
C1CCC(CC1)(C2=CC(=CC=C2)N=C=S)N3CCCCC3
300.464
300.46
Metaphit
C18H24N2S
FGSGBQAQSPSRJK-UHFFFAOYSA-N
1-[1-(3-isothiocyanatophenyl)cyclohexyl]piperidine
114745
>98%
Videos
Documents