missing translation for 'onlineSavingsMsg'
Learn More

Trifluralin-D14 in n-Nonane 100μg/mL, Fisher Chemical™

Catalog Number 12930334
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy
Concentration 100μg/mL
CAS 347841-79-6
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C13H16F3N3O4
InChI Key ZSDSQXJSNMTJDA-ZLKPZJALSA-N
SMILES CCCN(CCC)C1=C(C=C(C=C1[N+](=O)[O-])C(F)(F)F)[N+](=O)[O-]
IUPAC Name N,N-bis(1,1,2,2,3,3,3-heptadeuteriopropyl)-2,6-dinitro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol) 349.369
PubChem CID 102602132
Solvent or Matrix n-Nonane
Physical Form Liquid
Chemical Name or Material Trifluralin-D14
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'