missing translation for 'onlineSavingsMsg'
Learn More

Phenol D5 in Methanol 2000μg/mL, Fisher Chemical™

Catalog Number 12916103
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 2000μg/mL
CAS 4165-62-2
Packaging Ampoule
Quantity 1 mL
Type Organic Single-component Standard Solution
Molecular Formula C6H6O
MDL Number MFCD00084164
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
PubChem CID 12205419
Solvent or Matrix Methanol
Chemical Name or Material Phenol D5
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'