missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isodrin in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 100μg/mL |
CAS | 465-73-6 |
Packaging | Ampoule(s) |
Quantity | 1 mL |
Molecular Formula | C12H8Cl6 |
InChI Key | QBYJBZPUGVGKQQ-HSECHKQMSA-N |
SMILES | C1C2C=CC1C3C2C4(C(=C(C3(C4(Cl)Cl)Cl)Cl)Cl)Cl |
Molecular Weight (g/mol) | 364.896 |
PubChem CID | 102090390 |
Solvent or Matrix | Methanol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'