missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloridazon in Acetonitrile 10μg/mL, Fisher Chemical™
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 10μg/mL |
CAS | 1698-60-8 |
Packaging | Ampoule(s) |
Quantity | 10 mL |
Molecular Formula | C10H8ClN3O |
MDL Number | MFCD00055402 |
UN Number | 1648 |
InChI Key | WYKYKTKDBLFHCY-UHFFFAOYSA-N |
SMILES | NC1=C(Cl)C(=O)N(N=C1)C1=CC=CC=C1 |
IUPAC Name | 5-amino-4-chloro-2-phenylpyridazin-3-one |
Show More |
Safety and Handling
Product Identifier
- Standard solution Chloridazon,5-amino-4-chloro-2-phenylpyridazine-3-(2H)-one,pyrazon 10ug/ml in acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'