missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buprofezin in Cyclohexane 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
Specifications
Concentration | 100μg/mL |
CAS | 69327-76-0 |
Packaging | Ampoule |
Quantity | 1 mL |
Molecular Formula | C16H23N3OS |
UN Number | 1145 |
InChI Key | PRLVTUNWOQKEAI-UHFFFAOYSA-N |
SMILES | CC(C)N1C(=NC(C)(C)C)SCN(C1=O)C2=CC=CC=C2 |
IUPAC Name | 2-tert-butylimino-5-phenyl-3-propan-2-yl-1,3,5-thiadiazinan-4-one |
Molecular Weight (g/mol) | 305.44 |
Show More |
Safety and Handling
Product Identifier
- Standard solution Buprofezin 100ug/ml in cyclohexane
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P331-Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'