missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Met-Lys-Bradykinin

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J61532.MB

Code : NEW

Additional Details : CAS Number : 550-19-6 Weight : 0.00025kg

 View more versions of this product

Product Code. 15434119

  • £177.66 / 25mg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Specifications

1319.60
25mg
Met-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg
-30° to -10°C
HPLC
Powder
RUO
Met-Lys-Bradykinin
Videos
SDS

For Research Use Only