missing translation for 'onlineSavingsMsg'
Learn More

Mesulergine hydrochloride, Tocris Bioscience™

5-HT2A and 5-HT2C antagonist. Also dopamine receptor partial agonist

Brand:  Tocris (Bio-Techne) 164410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13459853

  • £175.56 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

5-HT2A and 2C receptor antagonist (pA2 values are 9.1 for each receptor) and D2-like dopamine receptor partial agonist (Ki = 8 nM). Displays antiprolactin and antiparkinsonian effects in vivo.
Specifications

Chemical Identifiers

72786-12-0
398.95
mesulergine hydrochloride, mesulergine hcl, n-1,6-dimethylergolin-8-alpha-yl-n,n-dimethylsulfamide hydrochloride, n'-8-alpha-1,6-dimethylergolin-8-yl-n,n-dimethylsulfamide monohydrochloride, sulfamide, n'-8-alpha-1,6-dimethylergolin-8-yl-n,n-dimethyl-, monohydrochloride, sulfamide, n'-8alpha-1,6-dimethylergolin-8-yl-n,n-dimethyl-, monohydrochloride, c18h26n4o2s.hcl, cq 32085 monohydrochloride, n'-8-?1,6-dimethylergolin-8-yl-n,n-dimethylsulfamide hydrochloride
(6aR,9S,10aR)-9-(dimethylsulfamoylamino)-4,7-dimethyl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline;hydrochloride
C18H27ClN4O2S
HANSYUJEPWNHIM-IVMONYBCSA-N
155746
CN1CC(CC2C1CC3=CN(C4=CC=CC2=C34)C)NS(=O)(=O)N(C)C.Cl

Specifications

Mesulergine hydrochloride
72786-12-0
mesulergine hydrochloride, mesulergine hcl, n-1,6-dimethylergolin-8-alpha-yl-n,n-dimethylsulfamide hydrochloride, n'-8-alpha-1,6-dimethylergolin-8-yl-n,n-dimethylsulfamide monohydrochloride, sulfamide, n'-8-alpha-1,6-dimethylergolin-8-yl-n,n-dimethyl-, monohydrochloride, sulfamide, n'-8alpha-1,6-dimethylergolin-8-yl-n,n-dimethyl-, monohydrochloride, c18h26n4o2s.hcl, cq 32085 monohydrochloride, n'-8-?1,6-dimethylergolin-8-yl-n,n-dimethylsulfamide hydrochloride
CN1CC(CC2C1CC3=CN(C4=CC=CC2=C34)C)NS(=O)(=O)N(C)C.Cl
398.95
398.95
10mg
C18H27ClN4O2S
HANSYUJEPWNHIM-IVMONYBCSA-N
(6aR,9S,10aR)-9-(dimethylsulfamoylamino)-4,7-dimethyl-6,6a,8,9,10,10a-hexahydroindolo[4,3-fg]quinoline;hydrochloride
155746
>99%
Videos