missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MESP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567086
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: ACPEGQAMEPS PPSPLLPGDV LALLE Highest antigen sequence identity to the following orthologs - mouse 65%, rat 62%.
MESP1 Polyclonal antibody specifically detects MESP1 in Human samples. It is validated for Immunocytochemistry.Specifications
MESP1 | |
Polyclonal | |
Unconjugated | |
MESP1 | |
bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; mesoderm posterior bHLH transcription factor 1; mesoderm posterior protein 1; MESP1; MGC10676 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRJ9 | |
MESP1 | |
Recombinant Human MESP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only