missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Mesothelioma Ab-1 Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelioma Ab-1, Mouse Monoclonal Antibody.
£97.67 - £364.00
Specifications
Antigen | Mesothelioma Ab-1 |
---|---|
Clone | HBME-1 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12619926
|
Epredia™
MS-1494-R7 |
Prediluted | 7 mL |
£209.00
7mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12649926
|
Epredia™
MS-1494-S1 |
Supernatant | 500 μL |
£265.00
500µL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12639926
|
Epredia™
MS-1494-S |
Supernatant | 1 mL |
£364.00
1mL |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12629926
|
Epredia™
MS-1494-RQ |
Ready-to-Use | 12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659926
|
Epredia™
MS-1494-S0 |
Supernatant | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Host Species: Mouse
Clone: HBME-1
Isotype: IgM/κ
Species Reactivity: Human. Others not known.
Immunogen: A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Mesothelioma Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma |
HBME-1 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Primary |