missing translation for 'onlineSavingsMsg'
Learn More

Mesoporphyrin IX dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7058822
Click to view available options
Quantity:
10 mg
50 mg
250 mg
This item is not returnable. View return policy
68938-72-7
C34H40Cl2N4O4
639.62
MFCD00136049
YUIUEEROGVTICN-UHFFFAOYSA-N
mesoporphyrin ix dihydrochloride, mesoporphyrin ix, dihydrochloride, 7,12-diethyl-3,8,13,17-tetramethyl-21h,23h-porphine-2,18-dipropionic acid dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, hydrochloride 1:2, mesoporphyrin ix dihydrochloride, synthetic, 3-18-2-carboxyethyl-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl propanoic acid dihydrochloride, 3-20-2-carboxyethyl-10,15-diethyl-5,9,14,19-tetramethyl-21,22,23,24-tetraazapentacyclo 16.2.1.1_,?.1?,__.1__,_? tetracosa-1,3 24 ,4,6,8,10,12,14,16 22 ,17,19-undecaen-4-yl propanoic acid dihydrochloride
111858
[H+].[H+].[Cl-].[Cl-].CCC1=C(C)C2=CC3=C(CC)C(C)=C(N3)C=C3N=C(C=C4N=C(C=C1N2)C(C)=C4CCC(O)=O)C(CCC(O)=O)=C3C
This item is not returnable. View return policy
68938-72-7
C34H40Cl2N4O4
639.62
MFCD00136049
YUIUEEROGVTICN-UHFFFAOYSA-N
mesoporphyrin ix dihydrochloride, mesoporphyrin ix, dihydrochloride, 7,12-diethyl-3,8,13,17-tetramethyl-21h,23h-porphine-2,18-dipropionic acid dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, dihydrochloride, 21h,23h-porphine-2,18-dipropanoic acid, 7,12-diethyl-3,8,13,17-tetramethyl-, hydrochloride 1:2, mesoporphyrin ix dihydrochloride, synthetic, 3-18-2-carboxyethyl-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl propanoic acid dihydrochloride, 3-20-2-carboxyethyl-10,15-diethyl-5,9,14,19-tetramethyl-21,22,23,24-tetraazapentacyclo 16.2.1.1_,?.1?,__.1__,_? tetracosa-1,3 24 ,4,6,8,10,12,14,16 22 ,17,19-undecaen-4-yl propanoic acid dihydrochloride
111858
[H+].[H+].[Cl-].[Cl-].CCC1=C(C)C2=CC3=C(CC)C(C)=C(N3)C=C3N=C(C=C4N=C(C=C1N2)C(C)=C4CCC(O)=O)C(CCC(O)=O)=C3C

Mesoporphyrin IX dihydrochloride is used reactant in constructing efficient dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 mg
Solubility Information Soluble in water.
IUPAC Name 3-[18-(2-carboxyethyl)-8,13-diethyl-3,7,12,17-tetramethyl-22,23-dihydroporphyrin-2-yl]propanoic acid;dihydrochloride
Formula Weight 639.62
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Mesoporphyrin IX dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'