missing translation for 'onlineSavingsMsg'
Learn More

meso-Erythritol, 99%, Thermo Scientific Chemicals

Catalog Number p-7022859
Click to view available options
Quantity:
25 g
100 g
149-32-6
C4H10O4
122.12
MFCD00004710
UNXHWFMMPAWVPI-UHFFFAOYNA-N
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
222285
CHEBI:17113
butane-1,2,3,4-tetrol
OCC(O)C(O)CO
149-32-6
C4H10O4
122.12
MFCD00004710
UNXHWFMMPAWVPI-UHFFFAOYNA-N
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
222285
CHEBI:17113
butane-1,2,3,4-tetrol
OCC(O)C(O)CO

meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

Solubility
Soluble in water (0.1 g/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at -20°C.
TRUSTED_SUSTAINABILITY
Melting Point 119°C to 124°C
Boiling Point 329°C to 331°C
Assay Percent Range 99%
Quantity 25 g
Beilstein 1719753
Merck Index 14,3675
Solubility Information Soluble in water (0.1g/ml).
Formula Weight 122.12
Percent Purity 99%
Chemical Name or Material meso-Erythritol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'