missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-4510050
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
608-36-6
C4H2Br2O4
273.87
MFCD00066439
FJWGRXKOBIVTFA-XIXRPRMCSA-L
2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
641611
(2S,3R)-2,3-dibromobutanedioic acid
[O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O
This item is not returnable. View return policy
608-36-6
C4H2Br2O4
273.87
MFCD00066439
FJWGRXKOBIVTFA-XIXRPRMCSA-L
2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
641611
(2S,3R)-2,3-dibromobutanedioic acid
[O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >275.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HO2CCH(Br)CH(Br)CO2H
Quantity 500 g
Beilstein 02,625
Merck Index 14,3029
Solubility Information Solubility in water: 20g/L (17°C). Other solubilities: soluble in alcohol and ether, sparingly soluble in chloroform
Formula Weight 275.88
Percent Purity 98%
Physical Form Powder
Chemical Name or Material meso-2,3-Dibromosuccinic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'