missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dibromosuccinic acid, 98%, ACROS Organics™

Brand:  Acros Organics 147570025

Code : 29

Additional Details : CAS Number : 608-36-6 Weight : 2.55000kg

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 11965031

  • £169.90 / 2.50kg


Estimated Shipment
13-03-2018
Add to basket

Specifications

Chemical Identifiers

meso-2,3-Dibromosuccinic acid
98%
HO2CCH(Br)CH(Br)CO2H
98%
C4H4Br2O4
275.88

Specifications

meso-2,3-Dibromosuccinic acid
> 275.0°C
608-36-6
Plastic bottle
98%
C4H4Br2O4
275.88
White to pale yellow to beige
2.5kg
Authentic
Powder
98%
HO2CCH(Br)CH(Br)CO2H
Solubility in water: 20g/L (17°C). Other solubilities: soluble in alcohol and ether, sparingly soluble in chloroform
Videos
SDS