missing translation for 'onlineSavingsMsg'
Learn More

Mesitylenesulfonic acid dihydrate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7028166
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
835617-36-2
C9H16O5S
236.282
MFCD00149535
LIKWMDGAWXCECN-UHFFFAOYSA-N
mesitylenesulfonic acid dihydrate, 2-mesitylenesulfonic acid dihydrate, 2,4,6-trimethylbenzenesulfonic acid dihydrate, benzenesulfonic acid, 2,4,6-trimethyl-, dihydrate, mesitylenesulfonicaciddihydrate, mesitylenesulfonic acid dihydrate, 98+%, benzenesulfonic acid,2,4,6-trimethyl-, hydrate 1:2, 2,4,6-trimethylbenzenesulfonic acid, oxamethane, oxamethane
15620344
2,4,6-trimethylbenzenesulfonic acid;dihydrate
CC1=CC(=C(C(=C1)C)S(=O)(=O)O)C.O.O
This item is not returnable. View return policy
835617-36-2
C9H16O5S
236.282
MFCD00149535
LIKWMDGAWXCECN-UHFFFAOYSA-N
mesitylenesulfonic acid dihydrate, 2-mesitylenesulfonic acid dihydrate, 2,4,6-trimethylbenzenesulfonic acid dihydrate, benzenesulfonic acid, 2,4,6-trimethyl-, dihydrate, mesitylenesulfonicaciddihydrate, mesitylenesulfonic acid dihydrate, 98+%, benzenesulfonic acid,2,4,6-trimethyl-, hydrate 1:2, 2,4,6-trimethylbenzenesulfonic acid, oxamethane, oxamethane
15620344
2,4,6-trimethylbenzenesulfonic acid;dihydrate
CC1=CC(=C(C(=C1)C)S(=O)(=O)O)C.O.O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 74°C to 77°C
Quantity 25 g
UN Number UN2585
Beilstein 1961501
Formula Weight 236.29 (200.26 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Mesitylenesulfonic acid dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'