missing translation for 'onlineSavingsMsg'
Learn More

Manutan Mesh blue door cabinet with three shelves, 1830mm x 915mm x 459mm

Brand:  Manutan A343047

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12872278

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Keeps stock secure, visible and ventilated. Welded 0. 9mm steel construction and reinforced mesh doors. Three-point locking lever handle with locks and keys supplied. Adjustable shelves.
  • Blue door
  • Three shelves
  • Dimensions (H x W x D): 1830mm x 915mm x 459mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers