missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MES, Monohydrate, Free Acidline, Crystal, Ultrapure Bioreagent, J.T. Baker™
419.90 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
200 g
Packaging:
Poly Bottle
Description
- For Liquid Chromatography and Molecular Biology Applications
Specifications
Packaging | Poly Bottle |
Quantity | 200 g |
Formula Weight | 213.25 |
Grade | Ultrapure Bioreagent |
Physical Form | Crystals |
Chemical Name or Material | MES, Monohydrate, Free Acidline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'