missing translation for 'onlineSavingsMsg'
Learn More

MES, Monohydrate, Free Acidline, Crystal, Ultrapure Bioreagent, J.T. Baker™

Catalog Number p-5391218
419.90 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
200 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
145224-94-8
C6H15NO5S
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
This item is not returnable. View return policy
145224-94-8
C6H15NO5S
213.248
MIIIXQJBDGSIKL-UHFFFAOYSA-N
mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology Applications
Packaging Poly Bottle
Quantity 200 g
Formula Weight 213.25
Grade Ultrapure Bioreagent
Physical Form Crystals
Chemical Name or Material MES, Monohydrate, Free Acidline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'