missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MES, 0.2M buffer soln., pH 6.0

Catalog Number 15423619
Click to view available options
Quantity:
250 mL
500 mL
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O
145224-94-8
C6H15NO5S
213.248
MFCD00283848
MIIIXQJBDGSIKL-UHFFFAOYSA-N
16218417
2-morpholin-4-ylethanesulfonic acid;hydrate
C1COCCN1CCS(=O)(=O)O.O

MES is used as a running buffer for resolving very small proteins (ex: 13 kDa) on bis-tris gels. Also used in cation-exchange chromatography and as a biological buffer in plant cell cultures. Used in various protein characterization assays. MES is also useful as a non-coordinating buffer in chemistry involving metal ions, as many common buffers (e.g. phosphate and acetate) readily form coordination complexes. MES only weakly binds Ca, Mg, Mn, and it has negligible binding with Cu(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a running buffer for resolving very small proteins (ex: 13 kDa) on bis-tris gels. Also used in cation-exchange chromatography and as a biological buffer in plant cell cultures. Used in various protein characterization assays. MES is also useful as a non-coordinating buffer in chemistry involving metal ions, as many common buffers (e.g. phosphate and acetate) readily form coordination complexes. MES only weakly binds Ca, Mg, Mn, and it has negligible binding with Cu(II).

Solubility
Soluble in water (∼1.3 M).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.2 M
pH 6.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water (∽1.3 M).
Chemical Name or Material MES buffer soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'