Learn More
MERTK Monoclonal Antibody (HMER5DS), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£190.00 - £386.00
Specifications
Antigen | MERTK |
---|---|
Clone | HMER5DS |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15993244
|
Affymetrix eBioscience
25-9043-41 |
25 Tests |
£190.00
Pack of 25 |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15903254
|
Affymetrix eBioscience
25-9043-42 |
100 Tests |
£386.00
Pack of 100 |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This HMER5DS antibody recognizes human MerTK, known also as Mer or c-mer, a 170-210 kDa member of the TAM family of tyrosine kinase receptors that also includes Axl and Tyro3. MerTK is expressed on a subset of anti-inflammatory macrophages and is involved in the removal of apoptotic cells. This process relies on two soluble ligands of Mer,TK Protein S and Gas6, that coat the surface of cells undergoing apoptosis. Upon binding these ligands, MerTK undergoes autophosphorylation at multiple tyrosine residues that activate the PI3K and Akt pathways. This results in the phagocytosis of target cells and also the direct inhibition of TLR-induced production of pro-inflammatory cytokines by the phagocytes. Mer may also function as a putative entry receptor for filoviruses. Deficiency of MerTK causes general autoimmunity, inflammation, and accumulation of apoptotic bodies. MerTK can be released from the cell surface by metalloproteinases, and this process is significantly enhanced when macrophages are stimulated, e.g. with LPS. MerTK is often expressed on maligt cells, and may be implicated in immune evasion. Applications Reported: This HMER5DS antibody has been reported for use in flow cytometric analysis. Applications Tested: This HMER5DS antibody has been pre-titrated and tested by flow cytometric analysis of human monocyte-derived macrophages. This can be used at 5 μL (0.06 μg) per test.
MERTK Monoclonal antibody specifically detects MERTK in Human samples. It is validated for Flow Cytometry.Specifications
MERTK | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
c-Eyk; c-mer; c-mer proto-oncogene tyrosine kinase; Eyk; MER; MER proto-oncogene, tyrosine kinase; MER receptor tyrosine kinase; MERTK; MGC133349; nmf12; Nyk; Proto-oncogene c-Mer; proto-oncogene tyrosine-protein kinase MER; rdy; receptor tyrosine kinase MerTK; Receptor tyrosine tinase gene probably the gene for Rdy; retinal dystrophy; RP38; sMER; sMERTK; soluble MER; soluble MERTK; STK kinase; Tyro 12; Tyro12; Tyrosine-protein kinase Mer | |
Mertk | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
HMER5DS | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
Q12866 | |
10461 | |
hMer-FC, PFA 1/2 + 1/2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Mertk |