missing translation for 'onlineSavingsMsg'
Learn More

MERTK, PE, clone: HMER5DS, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9043-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15587166

  • £88.05 / 25 tests


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This HMER5DS antibody recognizes human MerTK, known also as Mer or c-mer, a 170-210kDa member of the TAM family of tyrosine kinase receptors that also includes Axl and Tyro3. MerTK is expressed on a subset of anti-inflammatory macrophages and is involved in the removal of apoptotic cells. This process relies on two soluble ligands of Mer,TK Protein S and Gas6, that coat the surface of cells undergoing apoptosis. Upon binding these ligands, MerTK undergoes autophosphorylation at multiple tyrosine residues that activate the PI3K and Akt pathways. This results in the phagocytosis of target cells and also the direct inhibition of TLR-induced production of pro-inflammatory cytokines by the phagocytes. Mer may also function as a putative entry receptor for filoviruses. Deficiency of MerTK causes general autoimmunity, inflammation, and accumulation of apoptotic bodies. MerTK can be released from the cell surface by metalloproteinases, and this process is significantly enhanced when macrophages are stimulated, e.g. with LPS. MerTK is often expressed on malignant cells, and may be implicated in immune evasion.Applications Reported: This HMER5DS antibody has been reported for use in flow cytometric analysis.Applications Tested: This HMER5DS antibody has been pre-titrated and tested by flow cytometric analysis of human monocyte-derived macrophages. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

MerTK
HMER5DS
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Mer, c-mer
IgG1
RUO
Primary
Human
Videos