Learn More
Mercury Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Brand: Fisher Chemical J/8047/08
Code : 23
Additional Details : CAS Number : 7697-37-2 Weight : 0.51500kg Transport : UN number : 2031 Chem class : 8 Pack group : II
Quantity | 500mL |
---|
Chemical Identifiers
7783-34-8 | |
342.61 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+);dinitrate;hydrate |
H2HgN2O7 | |
MFCD00149736 | |
3084029 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
7783-34-8 | |
MFCD00149736 | |
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
342.61 | |
Liquid | |
1 | |
500mL |
H2HgN2O7 | |
KVICROHOONHSRH-UHFFFAOYSA-N | |
mercury(2+);dinitrate;hydrate | |
3084029 | |
HDPE plastic bottle | |
Colorless |