missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Mercury, plasma standard solution, Specpure™, Hg 10,000μg/mL

Brand:  Alfa Aesar™ 014395.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 100mL

 View more versions of this product

Product Code. 11352318

  • £67.60 / 100mL


Estimated Shipment date
from Supplier 22-05-2018
Add to basket

Specifications

Chemical Identifiers

Mercury plasma standard solution
Specpure™
UN3264
Hg 10000μg/mL
MFCD00011035

Specifications

Mercury plasma standard solution
Specpure™
Hg in 5% HNO3
UN3264
Hg 10000μg/mL
Liquid
MFCD00011035
100mL
Videos
SDS