missing translation for 'onlineSavingsMsg'
Learn More

Mercury, plasma standard solution, Specpure™ Hg 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4408569
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7439-97-6, 7697-37-2, 7732-18-5
Hg
200.59
MFCD00011035
QSHDDOUJBYECFT-UHFFFAOYSA-N
23931
CHEBI:16170
mercury
[Hg]
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7439-97-6, 7697-37-2, 7732-18-5
Hg
200.59
MFCD00011035
QSHDDOUJBYECFT-UHFFFAOYSA-N
23931
CHEBI:16170
mercury
[Hg]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 500 mL
Color Colorless
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Mercury: 0.1%; Water: 94.9%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Mercury

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'