Learn More
Mercury(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Hg(OTf)2 exhibits remarkable catalytic activity for the hydroxylative cyclization of 1,6-enynes. Mercuric triflate is a very versatile reagent and has been used for several organic catalytic transformations including C-C bond forming cyclizations, alkyne hydrations, heterocycle synthesis and very recently in C-N bond forming reactions. Commonly used as the metal source for Lewis acid catalyzed asymmetric reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >350°C |
Color | White |
Linear Formula | Hg(CF3SO3)2 |
Quantity | 1 g |
UN Number | UN2025 |
Sensitivity | Moisture sensitive |
Solubility Information | It is soluble in H2O and CH3CN,fairly soluble in CH3NO2 and CH2Cl2,insoluble in hexane,ether,and toluene. |
Formula Weight | 498.72 |
Percent Purity | 98% |
Physical Form | Powder |
Show More |
RUO – Research Use Only