missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Mercury(II) trifluoromethanesulfonate, 98%

Brand:  Alfa Aesar™ 039305.14

UNSPSC : 12352103

Code : NEW

Additional Details : CAS Number : 49540-00-3 Weight : 0.02500kg Transport : UN number : 2025 Chem class : 1 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11303718

  • £401.10 / 25g


Estimated Shipment date
from Supplier 19-09-2017
Add to basket

Specifications

Chemical Identifiers

Mercury(II) trifluoromethanesulfonate
Hg(CF3SO3)2
MFCD00144746
Mercury(II) triflate
49540-00-3
498.72
UN2025

Specifications

Mercury(II) trifluoromethanesulfonate
25g
White
Powder
Hg(CF3SO3)2
MFCD00144746
Mercury(II) triflate
>350°C
49540-00-3
Moisture Sensitive
98%
498.72
UN2025
Documentation

Technical Documents