Learn More
Mercury(II) trifluoroacetate, 98+%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Mercury(II) trifluoroacetate promotes selective cyclization of methylthio-substituted divinyl ketones to 5-fluorocyclopentenone derivatives. It is a raw material of fine chemicals and pharmaceutical intermediates. So it can be used in the pharmaceutical synthesis. Both linear and cross-linked copolymers of N-vinylcarbazole and methylacrylate can be prepared and mercuriated by using mercury trifluoroacetate in THF or dichloromethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 167°C to 169°C |
Color | White |
Linear Formula | Hg(OOCCF3)2 |
Quantity | 5 g |
UN Number | UN2025 |
Beilstein | 3633860 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 426.62 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only