missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Mercury(II) perchlorate hexahydrate, 99.999% (metals basis)

Brand:  Alfa Aesar™ 044317.30

Code : NEW

Additional Details : CAS Number : 7616-83-3 Weight : 0.25000kg Transport : UN number : 3087 Chem class : 1 Pack group : II

Quantity 250g

 View more versions of this product

Product Code. 11448420

  • £864.01 / 250g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Mercury (II) perchlorate hexahydrate
Cl2HgO8·6H2O
507.58 (399.49 Anhydrous)
UN3087
7616-83-3
Hg(ClO4)2·6H2O
MFCD00011036

Specifications

Mercury (II) perchlorate hexahydrate
Crystalline
(metals basis)
Hg(ClO4)2·6H2O
MFCD00011036
Off-white
7616-83-3
99.999%
Cl2HgO8·6H2O
507.58 (399.49 Anhydrous)
UN3087
250g
Videos
SDS