missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(II) oxide, yellow, ACS, 99.0% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Mercury(II) oxide production of mercury. It is also used as a material for cathodes for mercury batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 500°C (decomposition) |
Color | Orange |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.0% min. |
Solubility Information | Insoluble in water. Insoluble in alcohol,ether,acetone,ammonia. |
Formula Weight | 216.59 |
Grade | ACS |
Appearance | Dark orange |
Density | 11.14 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'