missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) nitrate dihydrate, ACS reagent, 9th edition, Thermo Scientific Chemicals

Catalog Number p-4440147
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
7782-86-7
Hg2N2O6·2H2O
561.22
This item is not returnable. View return policy
7782-86-7
Hg2N2O6·2H2O
561.22

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70.0°C
Quantity 50 g
Linear Formula Hg2(NO3)2·2H2O
Solubility Information Solubility in water: 20g/L (20°C)
Formula Weight 561.22
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Mercury(I) nitrate dihydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Mercury(I) nitrate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'