missing translation for 'onlineSavingsMsg'
Learn More

Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals

Catalog Number 12397193
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10112-91-1
Cl2Hg2
472.08
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
24956
CHEBI:33050
chloromercury
Cl[Hg].Cl[Hg]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10112-91-1
Cl2Hg2
472.08
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
mercurous chloride, calomel, mercury i chloride, mercury chloride hg2cl2, mercury subchloride, dimercury dichloride, calogreen, calotab, chlorure mercureux, ccris 3803
24956
CHEBI:33050
chloromercury
Cl[Hg].Cl[Hg]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 400.0°C
Quantity 100 g
Linear Formula Hg2Cl2
Merck Index 15,5962
Solubility Information Solubility in water: practically insoluble. Other solubilities: insoluble in ether and alcohol
Formula Weight 472.08
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Mercury(I) chloride
compliance-icons compliance-icons
Product Identifier
  • Mercury(I) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'