missing translation for 'onlineSavingsMsg'
Learn More

Mercury, 99.999%, (trace metal basis), ACROS Organics™

Brand:  Acros Organics 193480500

Code : 29

Additional Details : CAS Number : 7439-97-6 Weight : 0.05000kg Transport : UN number : 2809 Chem class : 1 Pack group : III

Quantity 50g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10636402

  • £23.50 / 50g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

Mercury
Trace Metal Basis
Hg
MFCD00011035
7439-97-6
99.999%
200.59
Quicksilver

Specifications

Mercury
7439-97-6
Trace Metal Basis
99.999%
200.59
09,291; 13,174; 15,198; 16,205
Quicksilver
0.001% max.
50g
13.5400g/mL
Heavy Liquid
Hg
MFCD00011035
15, 5966
Plastic bottle
Silver
Videos
SDS