missing translation for 'onlineSavingsMsg'
Learn More

Mercuric Acetate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390790
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
1600-27-7
C4H6HgO4
318.68
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
mercury 2+ ion acetic acid
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]
This item is not returnable. View return policy
1600-27-7
C4H6HgO4
318.68
BRMYZIKAHFEUFJ-UHFFFAOYSA-L
mercury 2+ ion acetic acid
15337
CHEBI:33211
mercury(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Hg+2]

High quality chemicals for laboratory and specialized industrial use

Physical Form Powder
Quantity 500 g
Formula Weight 318.68
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 3.27kg
Chemical Name or Material Mercuric Acetate
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'