missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Mercaptoethylamine Hydrochloride, Fisher BioReagents
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass
Description
β-Mercaptoethylamine HCl is used as an antioxidant in several biochemical procedures.
Specifications
Melting Point | 66°C |
Color | White |
pH | 3.3 to 5.0 |
Assay Percent Range | 98% |
Packaging | Amber Glass |
Quantity | 25 g |
Merck Index | 15, 2776 |
Formula Weight | 113.61 |
Percent Purity | ∽98% |
Physical Form | Solid |
Show More |
Safety and Handling
Product Identifier
- 2-Aminoethanethiol hydrochloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'