missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-7076539
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
367-51-1
C2H3NaO2S
114.094
MFCD00043386
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
23690444
CHEBI:86481
sodium;2-sulfanylacetate
C(C(=O)[O-])S.[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
367-51-1
C2H3NaO2S
114.094
MFCD00043386
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
23690444
CHEBI:86481
sodium;2-sulfanylacetate
C(C(=O)[O-])S.[Na+]

Reagent that protects tryptophan in amino acid analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Packaging Plastic bottle
Linear Formula HSCH2CO2Na
Quantity 25 g
UN Number UN2811
Beilstein 4569109
Sensitivity Air sensitive; Hygroscopic
Formula Weight 114.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mercaptoacetic acid sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'