missing translation for 'onlineSavingsMsg'
Learn More

Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-155688
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
367-51-1
C2H3NaO2S
114.09
MFCD00043386
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
23690444
CHEBI:86481
sodium;2-sulfanylacetate
C(C(=O)[O-])S.[Na+]
367-51-1
C2H3NaO2S
114.09
MFCD00043386
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate
23690444
CHEBI:86481
sodium;2-sulfanylacetate
C(C(=O)[O-])S.[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CO2Na
Quantity 50 g
Merck Index 15,8819
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: slightly soluble in alcohol
Formula Weight 114.09
Percent Purity 98%
Chemical Name or Material Mercaptoacetic acid, sodium salt, 98%
compliance-icons compliance-icons
Product Identifier
  • Mercaptoacetic acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'