Thermo Scientific™ MEPS™ for LC: Thermo Scientific, CTC Analytics, HTA and Varian 8400 Systems
Save hours in sample preparation, and go from extraction to injection in a single process with these 250µL syringes for LC applications using CTC Analytics.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: 60308-402
UNSPSC: 41104922
Code: 16
Additional Details:
Additional Details: Weight: 0.30000kg
Product Code. 10299164
Quantity Price
1 £ 261.0 / Pack of 5
Estimated Shipment
25-05-2017
Add to basket
Description and Specification
Specification
For Use With (Equipment) CTC Analytics, HTA, Varian 8400 Systems Using 100
Type MEPS Bin
Sorbent HyperSep Retain CX
Item Description Retain-CX MEPS for LC
For Use With CTC Analytics, HTA, Varian 8400 Systems Using 100
MEPS performs the same functions as SPE—the removal of interfering matrix components and the selective isolation and concentration of analytes—except that it miniaturizes conventional SPE packed bed devices from milliliter bed volumes to microliter volumes, and incorporates them right into the syringe.
Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC applications:
Significantly reduces the time to prepare and inject samples from hours to minutes—typically less than two minutes per sample
Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto GC
Reduces buffer and solvent volume from milliliters to microliters
Reduces the required sample volume to as little as 3.6µL, making the techniques especially well-suited for on-line use with volume-limited samples
Can easily be automated compatible and is compatible with CTC and eVol platforms