missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Meprin alpha Subunit/MEP1A Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | Meprin alpha Subunit/MEP1A |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
Meprin alpha Subunit/MEP1A Polyclonal specifically detects Meprin alpha Subunit/MEP1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
Meprin alpha Subunit/MEP1A | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
bA268F1.1 (meprin A alpha (PABA peptide hydrolase)), EC 3.4.24, EC 3.4.24.18, endopeptidase-2, MEP1A, meprin A subunit alpha, meprin A, alpha (PABA peptide hydrolase), N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha, PABA peptide hydrolase, PPH alpha, PPHA | |
MEP1A | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
AAA21338 | |
4224 | |
Mouse myeloma cell line NS0-derived recombinant human Meprin alpha Subunit/MEP1A Val22-Gln601 Accession # AAA21338 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Meprin alpha Subunit/MEP1A in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse MEP1A and recombinant human MEP1B is observed. |
For Research Use Only