missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melusin Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Melusin/ITGB1BP2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Melusin/ITGB1BP2 Polyclonal specifically detects Melusin/ITGB1BP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Melusin/ITGB1BP2 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CHORDC3, integrin beta 1 binding protein (melusin) 2, integrin beta-1-binding protein 2, ITGB1BP, ITGB1BP2, Melusin, MGC119214 | |
ITGB1BP2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q9UKP3 | |
26548 | |
E. coli-derived recombinant human Melusin Ser2-Glu347 Accession # Q9UKP3 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Melusin in direct ELISAs and Western blots. |
For Research Use Only