missing translation for 'onlineSavingsMsg'
Learn More

Melting Point Anthraquinone 283 to 286°C Standard, Reagecon

Ultrapure substance, specially prepared to calibrate, control, verify or qualify melting point (MP) devices.

Brand:  Reagecon RMP284

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13582100

  • £17.80 / 1g
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Melting Point Reference Standard, Melting Point Reference Material
  • The exceptional purity of this product is critical as MP will change with the presence of very small quantities of impurities.
  • This product is produced in accordance with the European, US and Chinese Pharmacopeia requirements.
  • The product can assist in identifying a compound or derivative or checking the purity or thermal stability of a known compound.
  • Tested and certified using a reference melting point apparatus that is calibrated using Certified Reference Materials to give traceability to the ITS-90 Temperature Scale.
  • Both the meniscus formation and complete liquefaction temperatures are certified.
Specifications

Specifications

Solid
1 x 1 g
Anthraquinone 283 to 286°C
Blue Glass Bottle
Accuracy ± 0.03°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers